Learn More
Medchemexpress LLC LC3-mHTT-IN-AN2 | 7758-73-8 | 98.1% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13025950MG
Encompass_Preferred
LC3-mHTT-IN-AN2 (Compound AN2) is a mHTT-LC3 linker compound. It interacts with both mutant huntingtin protein (mHTT) and LC3B, and reduces the levels of mHTT in an allele-selective manner in cultured Huntington disease (HD) mouse neurons.
  • Interacts with mutant huntingtin protein (mHTT) and LC3B
  • Reduces mutant huntingtin protein levels in an allele-selective manner
  • Supports research in Huntington disease (HD) mouse neurons
  • Target for autophagy and Atg8/LC3 pathway investigations
Catalog No. 50-003-39772