Learn More
Medchemexpress LLC LC3-mHTT-IN-AN2 | 7758-73-8 | 98.12% | 100 MG
SDP
Catalog No. 5000339897
Encompass_Preferred
LC3-mHTT-IN-AN2, also known as Compound AN2, is a mHTT-LC3 linker compound that interacts with both mutant huntingtin protein (mHTT) and LC3B, but not with wild-type HTT or irrelevant control proteins. It reduces the levels of mHTT in an allele-selective manner in cultured Huntington disease (HD) mouse neurons. This compound is for research use only.
  • Interacts with mutant huntingtin protein (mHTT) and LC3B
  • Does not interact with wild-type HTT or irrelevant control proteins
  • Reduces mHTT levels in an allele-selective manner
  • Useful in Huntington disease (HD) research
Catalog No. 50-003-39897 Supplier Medchemexpress LLC Supplier No. HY130259100MG
LC3-mHTT-IN-AN2, also known as Compound AN2, is a mHTT-LC3 linker compound that interacts with both mutant huntingtin protein (mHTT) and LC3B, but not with wild-type HTT or irrelevant control proteins. It reduces the levels of mHTT in an allele-selective manner in cultured Huntington disease (HD) mouse neurons. This compound is for research use only.
  • Interacts with mutant huntingtin protein (mHTT) and LC3B
  • Does not interact with wild-type HTT or irrelevant control proteins
  • Reduces mHTT levels in an allele-selective manner
  • Useful in Huntington disease (HD) research