Learn More
Sino Biological LC3A / MAP1LC3A Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 14322T44100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LC3A / MAP1LC3A (rh LC3A / MAP1LC3A; Catalog#14322-H08E; Q9H492-1; Met1-Phe121). LC3A / MAP1LC3A specific IgG was purified by Human LC3A / MAP1LC3A affinity chromatography.
Catalog No. 50-224-7668