Learn More
Sino Biological LC3B / MAP1LC3B Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 14555MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LC3B / MAP1LC3B (rh LC3B / MAP1LC3B; Catalog#14555-H07E; Q9GZQ8; Met1-Val125). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-224-8054