Learn More
Vector Laboratories Lens Culinaris Agglutinin (LCA), Unconjugated, 25mg
SDP
Catalog No. 502045667
Encompass
LCA recognizes sequences containing α-linked mannose residues but recognizes additional sugars as part of the receptor structure, giving it a narrower specificity than Con A. An α-linked fucose residue attached to the N-acetylchitobiose portion of the core oligosaccharide significantly enhances affinity. By exploiting this narrower specificity, glycoproteins and glycopeptides can be subfractionated with LCA after initial isolation with Con A.
Catalog No. 50-204-5667 Supplier Vector Laboratories Supplier No. L104025
Add to Cart
Add to Cart
LCA recognizes sequences containing α-linked mannose residues but recognizes additional sugars as part of the receptor structure, giving it a narrower specificity than Con A. An α-linked fucose residue attached to the N-acetylchitobiose portion of the core oligosaccharide significantly enhances affinity. By exploiting this narrower specificity, glycoproteins and glycopeptides can be subfractionated with LCA after initial isolation with Con A.