Learn More
Aviva Systems Biology Corp LCA5 Antibody - N-terminal region (ARP53410_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LCA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LCA5. Alternate names: C6orf152ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53715