Learn More
Medchemexpress LLC HY-120458 5mg , LCAHA CAS:117094-40-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1204585MG
Encompass_Preferred
HY-120458 5mg LCAHA CAS: 117094-40-3 LCAHA (LCA hydroxyamide) is a deubiquitinase USP2a inhibitor with IC50s of 9.7 μM and 3.7μM in Ub-AMC Assay and Di-Ub Assay, respectively. LCAHA destabilizes Cyclin D1 and induces G0/G1 arrest by inhibiting deubiquitinase USP2a. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1316