Learn More
Medchemexpress LLC Lcc-12 formic | 3029253-76-4 | 99.2% | 460.57 g/mol | C18H40N10O4 | 10 MG
SDP
Catalog No. 5000289330
Encompass_Preferred
LCC-12 formic is a research-grade small molecule probe (a dimer of metformin) used to modulate mitochondrial copper(II) and study inflammation and metabolic pathways in cellular and animal models.
  • Targets mitochondrial copper(II) and reduces the NAD(H) pool.
  • Reduces inflammation in mouse models of bacterial and viral infection.
  • Serves as a tool to study metabolic disease, cell plasticity, and epigenetic programming.
  • High purity and characterized molecular properties for reliable experimental use.
  • Compatible with storage at -20°C; in solution store at -80°C for long-term stability.
Catalog No. 50-002-89330 Supplier Medchemexpress LLC Supplier No. HY16793510MG
LCC-12 formic is a research-grade small molecule probe (a dimer of metformin) used to modulate mitochondrial copper(II) and study inflammation and metabolic pathways in cellular and animal models.
  • Targets mitochondrial copper(II) and reduces the NAD(H) pool.
  • Reduces inflammation in mouse models of bacterial and viral infection.
  • Serves as a tool to study metabolic disease, cell plasticity, and epigenetic programming.
  • High purity and characterized molecular properties for reliable experimental use.
  • Compatible with storage at -20°C; in solution store at -80°C for long-term stability.