Learn More
Medchemexpress LLC LCC-12 (formic) | 3029253-76-4 | 99.9% | 460.57 | 1 MG
SDP
Catalog No. 5000390670
Encompass_Preferred
LCC-12 formic is a dimer of metformin that targets mitochondrial copper(II), leading to a decrease in the NAD(H) pool and modulation of inflammatory responses. It reduces inflammation in mouse models of bacterial and viral infections and serves as a tool for investigating metabolic diseases through its effects on cell plasticity and epigenetic programming.
  • Targets mitochondrial copper(II)
  • Leads to a decrease in the NAD(H) pool
  • Modulates inflammatory responses
  • Reduces inflammation in mouse models of bacterial and viral infections
  • Serves as a tool for investigating metabolic diseases
  • Affects cell plasticity and epigenetic programming
Catalog No. 50-003-90670 Supplier Medchemexpress LLC Supplier No. HY1679351MG
LCC-12 formic is a dimer of metformin that targets mitochondrial copper(II), leading to a decrease in the NAD(H) pool and modulation of inflammatory responses. It reduces inflammation in mouse models of bacterial and viral infections and serves as a tool for investigating metabolic diseases through its effects on cell plasticity and epigenetic programming.
  • Targets mitochondrial copper(II)
  • Leads to a decrease in the NAD(H) pool
  • Modulates inflammatory responses
  • Reduces inflammation in mouse models of bacterial and viral infections
  • Serves as a tool for investigating metabolic diseases
  • Affects cell plasticity and epigenetic programming