Learn More
RDP CORPORATION LCC/LCD1-51 208 V 1- FULL POWR
Catalog No. NC3705253
Encompass
(LCC/LCD1-51)208 Volt/1-Phase Full PowerThe oven is designed to operate on 240 Volt, but will operate on 208 Volt without modifications, resulting in a 25% heater capacity reduction. 208 Volt operation without modifications requires 25% longer to reach the maximum operating temperature. The 208 Volt full power option restores the heater to full capacity. Lower voltage results in higher amperage. The FLA of the LCC/D1-51 with 208 Volt fullpower will increase from 27.7 amps to 31.5 amps. An oven equipped with the 208 Volt full power option cannot be operated on 240 Volt.
Catalog No. NC3705253 Supplier RDP CORPORATION Supplier No. 03.003 (LCC/LCD151)
(LCC/LCD1-51)208 Volt/1-Phase Full PowerThe oven is designed to operate on 240 Volt, but will operate on 208 Volt without modifications, resulting in a 25% heater capacity reduction. 208 Volt operation without modifications requires 25% longer to reach the maximum operating temperature. The 208 Volt full power option restores the heater to full capacity. Lower voltage results in higher amperage. The FLA of the LCC/D1-51 with 208 Volt fullpower will increase from 27.7 amps to 31.5 amps. An oven equipped with the 208 Volt full power option cannot be operated on 240 Volt.