Learn More
Research Products International Corp LCD Temperature/RH Recorder Set
SDP
Catalog No. 501977522
Encompass
These self-contained Relative Humidity (RH)/Temperature Dataloggers excel in the widest range of applications where regulatory compliance is required, food and drug production, shipping and storage, greenhouses, refrigerators, art galleries, museums, HVAC, process control, meteorology, agriculture, or labs. The scalability of this Datalogger provides for an unlimited number of additional Dataloggers being used by the same software.
Simply place the Datalogger in its docking station and program it using your computer and the Datalogger’s own software. You can set it to read temperature and RH in intervals of 1 second to 2 hours and for visual min/max alarms if desired. The flexibility of this Datalogger allows you to choose to begin recording immediately, at a predetermined time, or via external activation.
Set includes one Datalogger, docking station with computer cable, software, battery and instructions.
Catalog No. 50-197-7522 Supplier Research Products International Corp Supplier No. 800054
These self-contained Relative Humidity (RH)/Temperature Dataloggers excel in the widest range of applications where regulatory compliance is required, food and drug production, shipping and storage, greenhouses, refrigerators, art galleries, museums, HVAC, process control, meteorology, agriculture, or labs. The scalability of this Datalogger provides for an unlimited number of additional Dataloggers being used by the same software.
Simply place the Datalogger in its docking station and program it using your computer and the Datalogger’s own software. You can set it to read temperature and RH in intervals of 1 second to 2 hours and for visual min/max alarms if desired. The flexibility of this Datalogger allows you to choose to begin recording immediately, at a predetermined time, or via external activation.
Set includes one Datalogger, docking station with computer cable, software, battery and instructions.