Learn More
Waters Corp Vial, LCGC Certified, Amber Glass, 12 x 32 mm, 1.5 mL
Catalog No. 502390935 Shop All Waters Corporation Products
Encompass_Preferred
These LCGC Certified amber glass autosampler vials are designed for high-performance applications. They are tested for cleanliness by HPLC and manufactured to precise dimensional tolerances. This combination pack includes a vial, cap, and a PTFE septum, which is suitable for normal phase solvents but does not reseal after puncture.
  • Vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry.
  • Combination pack includes a PTFE septum for normal phase solvents.
  • Septum does not reseal after puncture.
Catalog No. 50-239-0935 Supplier Waters Corp Supplier No. 186007196C
These LCGC Certified amber glass autosampler vials are designed for high-performance applications. They are tested for cleanliness by HPLC and manufactured to precise dimensional tolerances. This combination pack includes a vial, cap, and a PTFE septum, which is suitable for normal phase solvents but does not reseal after puncture.
  • Vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry.
  • Combination pack includes a PTFE septum for normal phase solvents.
  • Septum does not reseal after puncture.