Learn More
Waters Corp Waters Corporation Screw Neck Vial, LCGC Certified Clear Glass, 12 x 32 mm, 2 mL Volume
Catalog No. 502037881 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters certified vials are HPLC-tested for cleanliness and manufactured to tight dimensional tolerances. They include a low bleed silicone/PTFE septum, ideal for normal phase solvents. Each batch is individually tested for chemical residues, and septa curing is verified by headspace GC. Made from chemically-inert glass, these vials prevent analytical interference. The PTFE/Silicone septum offers excellent resealing and supports a wide temperature range of –40 °C to 200 °C, ensuring sample integrity with its universal screw cap.
  • LCGC certified.
  • Screw neck closure.
  • Black cap.
  • 12 × 32 mm dimensions.
  • Clear glass.
  • PTFE/Silicone seal.
  • 2 mL volume.
  • Autosampler vial.
  • Includes vial, cap, septum.
  • For sample preparation.
  • 100 per package.
  • Septum temp range: –40 °C to 200 °C.
Catalog No. 50-203-7881 Supplier Waters Corp Supplier No. 186007199C
Waters certified vials are HPLC-tested for cleanliness and manufactured to tight dimensional tolerances. They include a low bleed silicone/PTFE septum, ideal for normal phase solvents. Each batch is individually tested for chemical residues, and septa curing is verified by headspace GC. Made from chemically-inert glass, these vials prevent analytical interference. The PTFE/Silicone septum offers excellent resealing and supports a wide temperature range of –40 °C to 200 °C, ensuring sample integrity with its universal screw cap.
  • LCGC certified.
  • Screw neck closure.
  • Black cap.
  • 12 × 32 mm dimensions.
  • Clear glass.
  • PTFE/Silicone seal.
  • 2 mL volume.
  • Autosampler vial.
  • Includes vial, cap, septum.
  • For sample preparation.
  • 100 per package.
  • Septum temp range: –40 °C to 200 °C.