Learn More
Waters Corp Waters Vial, LCGC Certified, Clear Glass, 12 x 32 mm, 1 mL
Catalog No. 502037885 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters LCGC Certified Clear Glass Total Recovery Vials are designed for optimal sample recovery and compatibility with both LC and GC techniques. These high-quality vials ensure durability, reliability, and secure sealing for sensitive analytical applications.
  • Tested for cleanliness by HPLC and held to tight dimensional tolerances.
  • Features a low bleed silicone/PTFE septum for normal phase solvents.
  • Total recovery design maximizes sample recovery and minimizes sample loss.
  • Compatible with LC and GC techniques for versatile analytical applications.
  • High-quality clear glass resists high temperatures and pressure.
  • Screw neck design ensures easy, secure sealing, preventing leakage or contamination.
  • Meets necessary standards and certifications for regulated environments.
Catalog No. 50-203-7885 Supplier Waters Corp Supplier No. 186007203C
Waters LCGC Certified Clear Glass Total Recovery Vials are designed for optimal sample recovery and compatibility with both LC and GC techniques. These high-quality vials ensure durability, reliability, and secure sealing for sensitive analytical applications.
  • Tested for cleanliness by HPLC and held to tight dimensional tolerances.
  • Features a low bleed silicone/PTFE septum for normal phase solvents.
  • Total recovery design maximizes sample recovery and minimizes sample loss.
  • Compatible with LC and GC techniques for versatile analytical applications.
  • High-quality clear glass resists high temperatures and pressure.
  • Screw neck design ensures easy, secure sealing, preventing leakage or contamination.
  • Meets necessary standards and certifications for regulated environments.