Learn More
Waters Corp Waters Corporation Vial, LCGC Certified, Clear Glass, 12 x 32 mm, 2 mL
Catalog No. 502037876 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters LCGC Certified Clear Glass Vials are rigorously tested for cleanliness and precision, ensuring reliable performance for sensitive analytical applications. This combination pack includes a PTFE septum, ideal for normal phase solvents, which ensures careful storage of samples.
  • HPLC tested for cleanliness; each batch examined.
  • Tightest dimensional tolerances ensure autosampler compatibility.
  • Chemically inert glass for accurate analytical results.
  • PTFE septum for normal phase solvents, suitable for many injections.
  • Septum offers excellent resealing, -40°C to 200°C operating range.
  • Universal screw cap prevents contamination during storage.
  • Certified to ISO 9001 dimensional standards.
Catalog No. 50-203-7876 Supplier Waters Corp Supplier No. 186007193C
Waters LCGC Certified Clear Glass Vials are rigorously tested for cleanliness and precision, ensuring reliable performance for sensitive analytical applications. This combination pack includes a PTFE septum, ideal for normal phase solvents, which ensures careful storage of samples.
  • HPLC tested for cleanliness; each batch examined.
  • Tightest dimensional tolerances ensure autosampler compatibility.
  • Chemically inert glass for accurate analytical results.
  • PTFE septum for normal phase solvents, suitable for many injections.
  • Septum offers excellent resealing, -40°C to 200°C operating range.
  • Universal screw cap prevents contamination during storage.
  • Certified to ISO 9001 dimensional standards.