Learn More
Enzo Life Sciences Lck (human), (recombinant) (GST-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3560005
Encompass_Preferred
Alternative name: p56lck. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 150mM NaCl, 0.25 mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol. Source: Produced in insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-1995