Learn More
Cayman Chemical LCK InhIbItr 500ug
SDP
Catalog No. 502970286
Encompass_Preferred
A pyrrolopyrimidine that selectively blocks the activity of two forms of the LCK kinase, LCK (64-509) and LCKCD with IC50 values of 50 = 4 and 25 mg/kg when administered either ip or orally)
Catalog No. 50-297-0286 Supplier Cayman Chemical Supplier No. 15135500 UG
Add to Cart
Add to Cart
A pyrrolopyrimidine that selectively blocks the activity of two forms of the LCK kinase, LCK (64-509) and LCKCD with IC50 values of 50 = 4 and 25 mg/kg when administered either ip or orally)