Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Lck, His Tag Recombinant Protein

Catalog No. 502740565
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-274-0565 10 μg
PR4170A 100 μg
PV4081 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-274-0565 Supplier Invitrogen™ Supplier No. P3043
Add to Cart
Edge
Add to Cart

Recombinant Protein

Specific Activity: 240 nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C.

LCK is a member of the Src-family tyrosine kinase, which are essential for signaling through the T-cell receptor (TCR) complex. Upon TCR triggering, LCK phosphorylates the ITAM motives in its zeta subunits establishing binding sites for the SH2 domains of the tyrosine kinase ZAP70, which is also phosphorylated by LCK and activated to generate subsequent signaling platforms by phosphorylation of adaptor LAT. The majority of LCK is localized to the plasma membrane, however, there is also a significant fraction associated with the Golgi apparatus which may contribute to Raf activation under conditions of weak stimulation through the TCR. LCK contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. LCK is also involved in the regulation of apoptosis induced by various stimuli, but not by the death receptors. Diseases associated with LCK include Immunodeficiency 22 and CD45 deficiency. Alternatively splice variants of the LCK gene encoding different isoforms of the protein have been found.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06239
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 3932
Molecular Weight (g/mol) 60.6 kDa
Name Human LCK, HIS Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C
Regulatory Status RUO
Gene Alias EC 2.7.10.2; Hck-3; IMD22; kinase Lck; Lck; LCK proto-oncogene, Src family tyrosine kinase; Lck1; Lcktkr; leukocyte C-terminal Src kinase; LSK; Lskt; Lsk-t; Lymphocyte cell-specific protein-tyrosine kinase; lymphocyte protein tyrosine kinase; lymphocyte-specific protein tyrosine kinase; OTTHUMP00000008640; OTTHUMP00000008740; OTTHUMP00000008741; p56(LSTRA) protein-tyrosine kinase; p56>lck>; p56lck; p56-LCK; pp58lck; Protein YT16; Proto-oncogene Lck; Proto-oncogene tyrosine-protein kinase LCK; RP4-675E8.4; t cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; tyr; tyrosine-protein kinase Lck; YT16
Common Name LCK
Gene Symbol LCK
Kinase Family SRC
Biological Activity 240 nM of phosphate transferred to poly [Glu, Tyr] 4:1 substrate / minute / mg of total protein at 30°C. Activity determined (at a final protein concentration of 2 μg/mL)
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Content And Storage -80°C
Activity 240 nM of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2 μg/mL
Expression System Insect cells
Protein Family Kinase and inhibitors
Form Liquid
Protein Subtype Cytoplasmic tyrosine kinase
Purity or Quality Grade 70% as determined by a SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.