Learn More
Medchemexpress LLC LCKLSL hydrochloride | 533902-29-3 | 98.05% | 712.34 | 25 MG
SDP
Catalog No. 5000426930
Encompass_Preferred
LCKLSL hydrochloride is a N-terminal hexapeptide and a competitive annexin A2 (AnxA2) inhibitor. It potently inhibits the binding of tissue plasminogen activator (tPA) to AnxA2. Additionally, this peptide has anti-angiogenic roles and inhibits the generation of plasmin.
  • N-terminal hexapeptide
  • Acts as a competitive annexin A2 (AnxA2) inhibitor
  • Potently inhibits the binding of tissue plasminogen activator (tPA) to AnxA2
  • Inhibits the generation of plasmin
  • Exhibits anti-angiogenic properties
  • Suppresses VEGF-induced tPA activity under hypoxic conditions
  • Decreases vascular length in angiogenesis models
Catalog No. 50-004-26930 Supplier Medchemexpress LLC Supplier No. 25MGHYP2333A
LCKLSL hydrochloride is a N-terminal hexapeptide and a competitive annexin A2 (AnxA2) inhibitor. It potently inhibits the binding of tissue plasminogen activator (tPA) to AnxA2. Additionally, this peptide has anti-angiogenic roles and inhibits the generation of plasmin.
  • N-terminal hexapeptide
  • Acts as a competitive annexin A2 (AnxA2) inhibitor
  • Potently inhibits the binding of tissue plasminogen activator (tPA) to AnxA2
  • Inhibits the generation of plasmin
  • Exhibits anti-angiogenic properties
  • Suppresses VEGF-induced tPA activity under hypoxic conditions
  • Decreases vascular length in angiogenesis models