Learn More
Medchemexpress LLC HY-P2333A 5mg Medchemexpress, LCKLSL (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP2333A5MG
Encompass_Preferred
Medchemexpress, HY-P2333A 5mg LCKLSL (hydrochloride) CAS: LCKLSL hydrochloride is a N-terminal hexapeptide and a competitive annexin A2 (AnxA2) inhibitor. LCKLSL hydrochloride potently inhibits the binding of tissue plasminogen activator (tPA) to AnxA2. LCKLSL hydrochloride also inhibits the generation of plasmin and has anti-angiogenic roles. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3086