Learn More
Medchemexpress LLC LCKLSL hydrochloride | 98.1% | 712.34 | 1 MG
SDP
Catalog No. 5000423559
Encompass_Preferred
LCKLSL hydrochloride is an N-terminal hexapeptide and a competitive annexin A2 (AnxA2) inhibitor. It potently inhibits the binding of tissue plasminogen activator (tPA) to AnxA2, inhibits the generation of plasmin, and has anti-angiogenic roles. In human retinal microvascular endothelial cells (RMVECs), LCKLSL inhibits plasmin generation and suppresses VEGF-induced tPA activity under hypoxic conditions. Application in in vivo models of angiogenesis demonstrates suppression of angiogenic responses.
  • Competitive annexin A2 (AnxA2) inhibitor
  • Inhibits binding of tissue plasminogen activator (tPA) to AnxA2
  • Inhibits generation of plasmin
  • Anti-angiogenic roles
  • Decreases vascular length in vivo
Catalog No. 50-004-23559 Supplier Medchemexpress LLC Supplier No. HYP2333A1MG
LCKLSL hydrochloride is an N-terminal hexapeptide and a competitive annexin A2 (AnxA2) inhibitor. It potently inhibits the binding of tissue plasminogen activator (tPA) to AnxA2, inhibits the generation of plasmin, and has anti-angiogenic roles. In human retinal microvascular endothelial cells (RMVECs), LCKLSL inhibits plasmin generation and suppresses VEGF-induced tPA activity under hypoxic conditions. Application in in vivo models of angiogenesis demonstrates suppression of angiogenic responses.
  • Competitive annexin A2 (AnxA2) inhibitor
  • Inhibits binding of tissue plasminogen activator (tPA) to AnxA2
  • Inhibits generation of plasmin
  • Anti-angiogenic roles
  • Decreases vascular length in vivo