Learn More
MSC Ridgid Vacuum Hose, Locking Pro, 1.875in Dia, 8ft, 1 ea
Catalog No. 502869497
Encompass
This Locking Pro wet/dry vacuum hose is engineered for industrial vacuum applications, offering durable plastic construction for reliable performance during various cleanup tasks.
  • Locking Pro hose type ensures secure attachment
  • Hose diameter of 1.875 in for efficient debris collection
  • Provides 8 ft hose length for extended reach
  • Constructed from durable plastic material
  • Gray color
  • Compatible with industrial wet/dry vacuums
Catalog No. 50-286-9497 Supplier MSC Supplier No. 46122487
This Locking Pro wet/dry vacuum hose is engineered for industrial vacuum applications, offering durable plastic construction for reliable performance during various cleanup tasks.
  • Locking Pro hose type ensures secure attachment
  • Hose diameter of 1.875 in for efficient debris collection
  • Provides 8 ft hose length for extended reach
  • Constructed from durable plastic material
  • Gray color
  • Compatible with industrial wet/dry vacuums