Learn More
Grainger Lckr 1x1Lckr Bl/Taupe 72 x12 x18 Assm
Catalog No. 502767168
Encompass_Preferred
The Grainger 16X110 Belt is a V-belt with a top width of 16mm and a length of 110cm. It is constructed of rubber with polyester cord reinforcement and has a type A cross-section profile. This V-belt is designed for use with small to medium pulley diameters and is commonly used in applications such as light-duty fans, pumps, and conveyors. The belt's rubber compound provides flexibility and friction grip while the polyester cords provide strength and prevent stretching. Overall, this is a quality V-belt well-suited for basic power transmission tasks involving small pulleys and lighter loads.
Catalog No. 50-276-7168 Supplier Grainger Supplier No. 16X110
The Grainger 16X110 Belt is a V-belt with a top width of 16mm and a length of 110cm. It is constructed of rubber with polyester cord reinforcement and has a type A cross-section profile. This V-belt is designed for use with small to medium pulley diameters and is commonly used in applications such as light-duty fans, pumps, and conveyors. The belt's rubber compound provides flexibility and friction grip while the polyester cords provide strength and prevent stretching. Overall, this is a quality V-belt well-suited for basic power transmission tasks involving small pulleys and lighter loads.