Learn More
Grainger Lckr 1x3Lckr Silver 78 x36 x18 UnAssm
Catalog No. 502751833
Encompass_Preferred
The Grainger 14L055 is a Westward brand 1/4-inch drive click-type torque wrench designed for accurate tightening and loosening of fasteners. It has a range of 10 to 150 inch pounds, featuring a knurled grip handle, satin chrome finish, and dual scale marked dial with a positive lock and release mechanism. This click-type wrench emits an audible click to alert when the pre-set torque value is reached. It can measure torque in clockwise and counterclockwise directions. The 14L055 click torque wrench is 15 inches long and weighs 1.4 lbs.
Catalog No. 50-275-1833 Supplier Grainger Supplier No. 14L055
The Grainger 14L055 is a Westward brand 1/4-inch drive click-type torque wrench designed for accurate tightening and loosening of fasteners. It has a range of 10 to 150 inch pounds, featuring a knurled grip handle, satin chrome finish, and dual scale marked dial with a positive lock and release mechanism. This click-type wrench emits an audible click to alert when the pre-set torque value is reached. It can measure torque in clockwise and counterclockwise directions. The 14L055 click torque wrench is 15 inches long and weighs 1.4 lbs.