Learn More
Grainger Lckr 1x3Lckr Silver 78 x36 x18 Assm
Catalog No. 502750274
Encompass_Preferred
The Grainger 14L067 is a Westward 3/8-inch drive deep impact socket set made in Taiwan. This 25-piece socket set includes sizes ranging from 4mm to 14mm, comes organized in a molded case, and has a lifetime replacement warranty. The chrome vanadium steel sockets feature a black phosphate finish for corrosion resistance.
Catalog No. 50-275-0274 Supplier Grainger Supplier No. 14L067
The Grainger 14L067 is a Westward 3/8-inch drive deep impact socket set made in Taiwan. This 25-piece socket set includes sizes ranging from 4mm to 14mm, comes organized in a molded case, and has a lifetime replacement warranty. The chrome vanadium steel sockets feature a black phosphate finish for corrosion resistance.