Learn More
Grainger Lckr 2x1Lckr Silver 78 x12 x18 UnAssm
Catalog No. 502751442
Encompass_Preferred
The Grainger Approved 2" x 1" Locker is a silver metal locker that measures 78" H x 12" W x 14" D. It features a recessed handle and 3-point locking mechanism with built-in combination lock. This locker provides a secure storage solution for personal belongings in various settings like schools, gyms, employee breakrooms and more. The all-welded steel construction ensures durability while the powder coat finish resists rust and corrosion. Overall, it's a high-quality locker that provides secure and reliable storage space.
Catalog No. 50-275-1442 Supplier Grainger Supplier No. 14L058
The Grainger Approved 2" x 1" Locker is a silver metal locker that measures 78" H x 12" W x 14" D. It features a recessed handle and 3-point locking mechanism with built-in combination lock. This locker provides a secure storage solution for personal belongings in various settings like schools, gyms, employee breakrooms and more. The all-welded steel construction ensures durability while the powder coat finish resists rust and corrosion. Overall, it's a high-quality locker that provides secure and reliable storage space.