Learn More
Grainger Lckr 3x1Lckr Taupe 72 x12 x18 Assm
Catalog No. 502766962
Encompass_Preferred
The Grainger 16X116 is a 16 inch x 116 inch, 0.006 inch thick clear polyethylene sheet. It is transparent and flexible with good chemical resistance. The material is LDPE, low density polyethylene. It has a maximum continuous service temperature of 180°F and a minimum of -70°F. Typical applications include dust covers, packaging, surface protection, and weather barriers. It meets FDA regulations for food contact. The sheet can be cut, folded, or seamed. It is sold by the each.
Catalog No. 50-276-6962 Supplier Grainger Supplier No. 16X116
The Grainger 16X116 is a 16 inch x 116 inch, 0.006 inch thick clear polyethylene sheet. It is transparent and flexible with good chemical resistance. The material is LDPE, low density polyethylene. It has a maximum continuous service temperature of 180°F and a minimum of -70°F. Typical applications include dust covers, packaging, surface protection, and weather barriers. It meets FDA regulations for food contact. The sheet can be cut, folded, or seamed. It is sold by the each.