Learn More
Cayman Chemical LCL-161phenol 10mg
SDP
Catalog No. 502945535
Encompass_Preferred
A derivative of LCL-161 and a PROTAC synthetic intermediate; has been used in the synthesis of PROTACs that drive degradation of androgen receptors
Catalog No. 50-294-5535 Supplier Cayman Chemical Supplier No. 4035810 MG
A derivative of LCL-161 and a PROTAC synthetic intermediate; has been used in the synthesis of PROTACs that drive degradation of androgen receptors