Learn More
Cayman Chemical LCL-161, phenol, Purity: ≥98%, CAS: 2095244-42-9, 50 mg
SDP
Catalog No. 502943320
Encompass_Preferred
A derivative of LCL-161 and a PROTAC synthetic intermediate; has been used in the synthesis of PROTACs that drive degradation of androgen receptors
Catalog No. 50-294-3320 Supplier Cayman Chemical Supplier No. 4035850 MG
A derivative of LCL-161 and a PROTAC synthetic intermediate; has been used in the synthesis of PROTACs that drive degradation of androgen receptors