Learn More
TARGETMOL CHEMICALS INC LCL161 10MG
SDP
Catalog No. 502577232
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LCL161 a SMAC mimetic effectivity binds to and inhibits multiple IAPs (i.e. XIAP c-IAP). purity: 99%
Catalog No. 50-257-7232 Supplier TARGETMOL CHEMICALS INC Supplier No. T208010MG
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LCL161 a SMAC mimetic effectivity binds to and inhibits multiple IAPs (i.e. XIAP c-IAP). purity: 99%