Learn More
Medchemexpress LLC HY-15518 100mg Medchemexpress, LCL161 CAS:1005342-46-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15518100MG
Encompass_Preferred
Medchemexpress,HY-15518 100mg LCL161 CAS:1005342-46-0 Formula:C26H33FN4O3S LCL161 is a IAP inhibitor which inhibits XIAP in HEK293 cell and cIAP1 in MDA-MB-231 cell with IC50s of 35 and 0.4 nM, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8327