Learn More
Abcam LCL161, 100MG
Catalog No. 502682918
Encompass_Preferred
MW 500.6 Da, Purity >99%. Small molecule SMAC mimetic inhibitor of multiple IAP (Inhibitor of apoptosis) family proteins. (IC₅₀ = 10.23 μM). Inhibits cell proliferation and viability in two human hepatocellular carcinoma cells (Hep3B, IC₅₀ = 10, and PLC5, IC₅₀ = 19).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2918 Supplier Abcam Supplier No. AB273616100MG
MW 500.6 Da, Purity >99%. Small molecule SMAC mimetic inhibitor of multiple IAP (Inhibitor of apoptosis) family proteins. (IC₅₀ = 10.23 μM). Inhibits cell proliferation and viability in two human hepatocellular carcinoma cells (Hep3B, IC₅₀ = 10, and PLC5, IC₅₀ = 19).
The product is subject to the following: Abcam Restricted Use Statement