Learn More
Apexbio Technology LLC LCL161 1005342-46-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A35415.1
Encompass_Preferred
LCL161 (CAS 1005342-46-0) is a small-molecule antagonist targeting inhibitor of apoptosis (IAP) proteins specifically designed as a mimetic of the endogenous second mitochondria-derived activator of caspase (SMAC) LCL161 binds to and inhibits XIAP and cIAP1/2 proteins frequently overexpressed in malignant cells It demonstrates antiproliferative activity in vitro against selected hepatocellular carcinoma cell lines (Hep3B PLC5) and hematologic cancer models (ALL) with IC50 values in the micromolar range In in vivo tumor xenograft studies oral administration of LCL161 has led to significant growth delays across various malignancies including osteosarcoma and glioblastoma Its combination with AAVP-delivered TNF- results in synergistic antitumor effects indicating its utility in oncology research
Catalog No. 50-000-64164