Learn More
Proteochem Inc LCMS Acetonitrile 4 x 4 L
Catalog No. NC3106240
Encompass_Preferred
LCMS Acetonitrile is shipped as a dangerous good (HazMat), UN1648 (Flammable Liquid, Product Group II, Hazard Class 3, Freight Class 65), and is packed as four 4L bottles in a 15"x15"x17" case weighing 39 lbs.
Catalog No. NC3106240 Supplier Proteochem Inc Supplier No. LC63104X4L
LCMS Acetonitrile is shipped as a dangerous good (HazMat), UN1648 (Flammable Liquid, Product Group II, Hazard Class 3, Freight Class 65), and is packed as four 4L bottles in a 15"x15"x17" case weighing 39 lbs.