Learn More
Proteochem Inc LCMS Acetonitrile with 0.1% Formic Acid and 0.01% TFA v/v
Catalog No. NC2467772
Encompass_Preferred
LCMS Acetonitrile with 0.1% Formic Acid and 0.01% TFA v/v
Catalog No. NC2467772 Supplier Proteochem Inc Supplier No. LC63141L
LCMS Acetonitrile with 0.1% Formic Acid and 0.01% TFA v/v