Learn More
Covachem Triethylamine, LC/MS, TEA, Proteomics, 121-44-8
Catalog No. NC1417016
Encompass
Triethylamine is a clear, colorless liquid with a fishy odor. It is used as a solvent, catalyst, and base in organic synthesis and other chemical processes. The chemical formula is N(CH2CH3)3 and molecular weight is 101.19 g/mol. It has a boiling point of 89°C, melting point of -115°C, density of 0.726 g/mL at 20°C, and is miscible with water and most organic solvents. Triethylamine is classified as a flammable liquid and corrosive chemical. It should be handled with care while wearing proper PPE. The product is available from CovaChem in various high purity grades suitable for analytical, biopharmaceutical, and other applications.
Catalog No. NC1417016 Supplier Covachem Supplier No. 1120525
Triethylamine is a clear, colorless liquid with a fishy odor. It is used as a solvent, catalyst, and base in organic synthesis and other chemical processes. The chemical formula is N(CH2CH3)3 and molecular weight is 101.19 g/mol. It has a boiling point of 89°C, melting point of -115°C, density of 0.726 g/mL at 20°C, and is miscible with water and most organic solvents. Triethylamine is classified as a flammable liquid and corrosive chemical. It should be handled with care while wearing proper PPE. The product is available from CovaChem in various high purity grades suitable for analytical, biopharmaceutical, and other applications.