Learn More
Covachem LCMS GRD FORMC ACID ULTPR 25ML
Catalog No. NC0922293
Encompass
LCMS Grade Formic Acid (UltraPure). 25 mL Bottle. Appearance: Clear liquid. Purity: is ≥ 99%. Storage: Room Temperature. 64-18-6.
Catalog No. NC0922293 Supplier Covachem Supplier No. 1120225
LCMS Grade Formic Acid (UltraPure). 25 mL Bottle. Appearance: Clear liquid. Purity: is ≥ 99%. Storage: Room Temperature. 64-18-6.