Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION LCMS QC Reference Standard, Reversed Phase, MS Detection, Liquid, 500 μL

Catalog No. 50596916 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The LCMS QC Reference Material is a 9-component mix containing Acetaminophen, Caffeine, Sulfaguanidine, Sulfadimethoxine, Val,-Tyr-Val, Verapamil, Terfenadine, Leucine-Enkephalin, and Reserpine, packaged in a specialized vial. This standard provides a comprehensive reference for use with LC/MS or MS instrumentation across a wide variety of conditions and methods.

  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Liquid
  • Storage condition: 4°C (fridge)
  • System type: LC, LCMS
  • Quantitative: Yes
  • Shelf life: 12 months
  • Shipping condition: Ambient
  • Volume: 500 μL
  • Type: Reference
  • Application: Small molecule
  • Product type: QC
  • Units per package: 1 pk

Catalog No. 50-596-916 Supplier Waters Corp Supplier No. 186006963
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The LCMS QC Reference Material is a 9-component mix containing Acetaminophen, Caffeine, Sulfaguanidine, Sulfadimethoxine, Val,-Tyr-Val, Verapamil, Terfenadine, Leucine-Enkephalin, and Reserpine, packaged in a specialized vial. This standard provides a comprehensive reference for use with LC/MS or MS instrumentation across a wide variety of conditions and methods.

  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Liquid
  • Storage condition: 4°C (fridge)
  • System type: LC, LCMS
  • Quantitative: Yes
  • Shelf life: 12 months
  • Shipping condition: Ambient
  • Volume: 500 μL
  • Type: Reference
  • Application: Small molecule
  • Product type: QC
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.