Learn More
Sino Biological Recombinant Human KAT7/Hbo1/MYST2 Protein (Full Length, K277Q, GST Tag), Active
Catalog No. 502574523
Encompass_Preferred
Recombinant full length human KAT7 (K277Q) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4523 Supplier Sino Biological Supplier No. K316382AG50
Recombinant full length human KAT7 (K277Q) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.