Learn More
Medchemexpress LLC LCMV GP (61-80) | 232598-19-5 | 99.6% | 2290.60 | 5 MG
SDP
Catalog No. 5000423298
Encompass_Preferred
LCMV GP (61-80) is a peptide fragment derived from the lymphocytic choriomeningitis virus (LCMV) glycoprotein (GP), corresponding to amino acids 61-80. It is a specific epitope that can induce a CD4+ T-cell response.
  • Stimulates splenocytes for detection of CD4 T cells in the spleen
  • Induces the proliferation of antigen-specific CD4 memory T cells in the bone marrow
Catalog No. 50-004-23298 Supplier Medchemexpress LLC Supplier No. HYP25605MG
LCMV GP (61-80) is a peptide fragment derived from the lymphocytic choriomeningitis virus (LCMV) glycoprotein (GP), corresponding to amino acids 61-80. It is a specific epitope that can induce a CD4+ T-cell response.
  • Stimulates splenocytes for detection of CD4 T cells in the spleen
  • Induces the proliferation of antigen-specific CD4 memory T cells in the bone marrow