Learn More
Medchemexpress LLC LCMV GP (61-80) | 232598-19-5 | 99.8% | 2290.60 | 10 MG
SDP
Catalog No. 5000423296
Encompass_Preferred
LCMV GP (61-80) is a peptide fragment derived from lymphocytic choriomeningitis virus (LCMV) glycoprotein (GP), corresponding to amino acids 61-80. This specific epitope can induce a CD4+ T-cell response and is intended for research use only.
  • Stimulates splenocytes for detection of LCMV-specific CD4 T cells in the spleen.
  • Induces proliferation of antigen-specific CD4 memory T cells in bone marrow of C57BL/6 mice.
Catalog No. 50-004-23296 Supplier Medchemexpress LLC Supplier No. HYP256010MG
LCMV GP (61-80) is a peptide fragment derived from lymphocytic choriomeningitis virus (LCMV) glycoprotein (GP), corresponding to amino acids 61-80. This specific epitope can induce a CD4+ T-cell response and is intended for research use only.
  • Stimulates splenocytes for detection of LCMV-specific CD4 T cells in the spleen.
  • Induces proliferation of antigen-specific CD4 memory T cells in bone marrow of C57BL/6 mice.