Learn More
Medchemexpress LLC HY-P1569 1mg Medchemexpress, LCMV gp33-41 CAS:151705-84-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP15691MG
Encompass_Preferred
Medchemexpress, HY-P1569 1mg LCMV gp33-41 CAS:151705-84-9 LCMV gp33-41, the carboxyl-extended 11-aa-long peptide, is an lymphocytic choriomeningitis virus sequence restricted by MHC class I H-2Db molecules and presented to cytotoxic T lymphocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1691