Learn More
Aviva Systems Biology Corp LCN12 Antibody - N-terminal region (ARP58489_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58489_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LCN12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LCN12. Alternate names: MGC34753, MGC48935ApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-62323