Learn More
Aviva Systems Biology Corp LCN8 Antibody - N-terminal region (ARP58490_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58490_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LCN8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LCN8. Alternate names: EP17, LCN5ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62324