Learn More
Aviva Systems Biology Corp LCOR Antibody - middle region (ARP59845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LCOR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LCOR. Alternate names: FLJ38026, KIAA1795, MLR2, RP11-175O19.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-852