Learn More
Aviva Systems Biology Corp Lcorl Antibody - middle region (ARP40132_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40132_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lcorl. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Mlr1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-854