Learn More
Hach Company LCP to pHD retrofit Union Mnt
Catalog No. 5000207086 Shop All Hach Company Products
Encompass
LCP To pHD Retrofit Union Mounting Hardware Adapter Kit For Existing 1 1/2 Inch Npt Tee For Epoxy Sensor (6051P Style) Cpvc Adapter Kit
Catalog No. 50-002-07086 Supplier Hach Company Supplier No. 60A2100001
LCP To pHD Retrofit Union Mounting Hardware Adapter Kit For Existing 1 1/2 Inch Npt Tee For Epoxy Sensor (6051P Style) Cpvc Adapter Kit