Learn More
Hach Company LCP Salt Bridge-Cer/Cer-pk/1
Supplier:  Hach Company 609765010001
Encompass
Field replaceable salt bridge has a ceramic inner junction ceramic outer junction FKM/FPM O-Ring and contains binary fill solution Only for use with LCP 1 5in differential pH sensors
Catalog No. 50-002-07210