Learn More
Aviva Systems Biology Corp LCP1 Antibody - N-terminal region (ARP54686_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54686_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LCP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LCP1. Alternate names: CP64, DKFZp781A23186, FLJ25423, FLJ26114, FLJ39956, L-PLASTIN, LC64P, PLS2, LPLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61284