Learn More
Cole-Parmer Amprobe LCR55A LCR Tester with Transistor Test
Supplier: Cole-Parmer 2002145
The MNCR50 is a pocket-sized capacitance and resistance meter that measures capacitance in 9 ranges from 200pF to 20mF; seven ranges for resistance from 20 ohm to 20Mohm. The MNLCR55 measures inductance from 200uH to 200H; capacitance measures ranges from 200pF to 20mF, and resistance from 20ohm to 20Mohm.